Lactose octaacetate, 98%
Lactose octaacetate, 98%
Thermo Scientific Chemicals

Lactose octaacetate, 98%

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number J66695.06
also known as J66695-06
Price (USD)/ Each
37.20
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
37.20
Add to cart
Lactose octaacetate, 98%
Catalog numberJ66695.06
Price (USD)/ Each
37.20
-
Add to cart
Chemical Identifiers
CAS6291-42-5
IUPAC Name[4,5,6-tris(acetyloxy)-3-{[3,4,5-tris(acetyloxy)-6-[(acetyloxy)methyl]oxan-2-yl]oxy}oxan-2-yl]methyl acetate
Molecular FormulaC28H38O19
InChI KeyWOTQVEKSRLZRSX-UHFFFAOYNA-N
SMILESCC(=O)OCC1OC(OC(C)=O)C(OC(C)=O)C(OC(C)=O)C1OC1OC(COC(C)=O)C(OC(C)=O)C(OC(C)=O)C1OC(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Proton NMRConforms to structure
Purity≥97.5%
FormPowder
Lactose octaacetate is used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Lactose octaacetate is used as pharmaceutical intermediates.

Solubility
Soluble in chloroform

Notes
Protect from heat. Incompatible with heat and oxidizing agents.
RUO – Research Use Only
L. P. Kuhn. Infrared Spectra of Carbohydrates. Anal. Chem.1950, 22, (2), 276-283.