Thermo Scientific Chemicals

Picloram, 95%, Thermo Scientific Chemicals

Catalog number: J66702.14
25 g, Each
Thermo Scientific Chemicals

Picloram, 95%, Thermo Scientific Chemicals

Catalog number: J66702.14
25 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: J66702.14
also known as J66702-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1918-02-1
IUPAC Name
4-amino-3,5,6-trichloropyridine-2-carboxylic acid
Molecular Formula
C6H3Cl3N2O2
InChI Key
NQQVFXUMIDALNH-UHFFFAOYSA-N
SMILES
NC1=C(Cl)C(Cl)=NC(C(O)=O)=C1Cl
Appearance (Color)
White to pale yellow to pale cream
Assay from Supplier's CofA
≥94.0%
Water Content
≤1.0%
Form
Powder

Description

Belongs to the puridine class of the auxinic compounds similar to the hormone Indole-3-acetic acid (IAA)

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text