Agarose, high gelling temperature, Molecular Biology Reagent
Agarose, high gelling temperature, Molecular Biology Reagent
Thermo Scientific Chemicals

Agarose, high gelling temperature, Molecular Biology Reagent

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number J66704.14
also known as J66704-14
Price (USD)/ Each
173.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
173.00
Add to cart
Agarose, high gelling temperature, Molecular Biology Reagent
Catalog numberJ66704.14
Price (USD)/ Each
173.00
-
Add to cart
Chemical Identifiers
CAS9012-36-6
IUPAC Name(2S,3R,4S,5R,6R)-2-{[(1S,3S,4S,5S,8R)-3-{[(2S,3R,4S,5S,6R)-2-{[(1S,3R,4S,5S,8R)-3,4-dihydroxy-2,6-dioxabicyclo[3.2.1]octan-8-yl]oxy}-3,5-dihydroxy-6-(hydroxymethyl)oxan-4-yl]oxy}-4-hydroxy-2,6-dioxabicyclo[3.2.1]octan-8-yl]oxy}-6-(hydroxymethyl)oxane-3,4,5-triol
Molecular FormulaC24H38O19
InChI KeyMJQHZNBUODTQTK-WKGBVCLCSA-N
SMILESOC[C@H]1O[C@@H](O[C@@H]2[C@@H]3CO[C@H]2[C@H](O)[C@H](O[C@H]2[C@@H](O)[C@@H](CO)O[C@@H](O[C@@H]4[C@@H]5CO[C@H]4[C@H](O)[C@H](O)O5)[C@@H]2O)O3)[C@H](O)[C@@H](O)[C@H]1O
View more
SpecificationsSpecification SheetSpecification Sheet
CommentElectroendosmosis, Mr: 0.09-0.13
Gel strengthGel Strength at 1%: ≥800g/cm2
CommentGelling temperature: 42.0 ±1.5?C
Water Content (Karl Fischer Titration)≤10.0%
High gel strength allows gel formation at low concentrations. Ideal for antibiotic sensitivity assays, RID, double diffusion and other immunological applications

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • High gel strength allows gel formation at low concentrations
  • Ideal for antibiotic sensitivity assays, RID, double diffusion and other immunological applications
RUO – Research Use Only