D-myo-Inositol-1,2,6-trisphosphate sodium salt, 98%, Thermo Scientific Chemicals
D-myo-Inositol-1,2,6-trisphosphate sodium salt, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

D-myo-Inositol-1,2,6-trisphosphate sodium salt, 98%, Thermo Scientific Chemicals

Have Questions?
Quantity:
0.1 mg
Catalog number J66721.EXZ
also known as J66721-!Z
Price (USD)/ Each
96.20
-
Add to cart
Quantity:
0.1 mg
Request bulk or custom format
Price (USD)/ Each
96.20
Add to cart
D-myo-Inositol-1,2,6-trisphosphate sodium salt, 98%, Thermo Scientific Chemicals
Catalog numberJ66721.EXZ
Price (USD)/ Each
96.20
-
Add to cart
Specifications
Chemical Name or MaterialD-myo-Inositol-1,2,6-trisphosphate sodium salt
Storage Note 1-30° to -10°C
ColorWhite
Recommended StorageStore at -20°C
TSCANo
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only