[Lys<sup>8</sup>] Vasopressin
Thermo Scientific Chemicals

[Lys8] Vasopressin

 
Have Questions?
Quantity:
1 mg
Catalog number J66728.MCR
also known as J66728-M^
Price (USD)/ Each
65.70
-
Add to cart
Quantity:
1 mg
Request bulk or custom format
Price (USD)/ Each
65.70
Add to cart
[Lys8] Vasopressin
Catalog numberJ66728.MCR
Price (USD)/ Each
65.70
-
Add to cart
SpecificationsSpecification SheetSpecification Sheet
Purification MethodHPLC
FormatSolid
Molecular Weight (g/mol)1056.22
Product Type[Lys8] Vasopressin
Quantity1 mg
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Contents & Storage
-30°C to -10°C