Thermo Scientific Chemicals
Sulfamonomethoxine, 98%, Thermo Scientific Chemicals
Catalog number: J66733.03
1 g, Each
Thermo Scientific Chemicals
Sulfamonomethoxine, 98%, Thermo Scientific Chemicals
Catalog number: J66733.03
1 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: J66733.03
also known as J66733-03
Price (USD)
Quantity
-
Have Questions?
Description
Sulfamonomethoxine is a long acting sulfonamide antibacterial agent. It is used to study degradation by Fe3O4 magnetic nanoparticles. It is used in blood kinetic studies and is used to study capsule formation of Bordetella bronchiseptica.