Cefamandole nafate, Thermo Scientific Chemicals
Cefamandole nafate, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Cefamandole nafate, Thermo Scientific Chemicals

A second generation cephalosporin antibiotic
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
5 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J66735.09
also known as J66735-09
Price (USD)/ Each
302.65
Online exclusive
336.00 
Save 33.35 (10%)
-
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)/ Each
302.65
Online exclusive
336.00 
Save 33.35 (10%)
Add to cart
Cefamandole nafate, Thermo Scientific Chemicals
Catalog numberJ66735.09
Price (USD)/ Each
302.65
Online exclusive
336.00 
Save 33.35 (10%)
-
Add to cart
Specifications
Chemical Name or MaterialCefamandole nafate
TypeCefamandole Nafate
Physical FormPowder
CAS42540-40-9
Health Hazard 1H315-H317-H319-H334-H335
View more
A second generation cephalosporin antibiotic.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only