Thermo Scientific Chemicals

Sucralose, 98%

Catalog number: J66736.18
50 g, Each
Thermo Scientific Chemicals

Sucralose, 98%

Catalog number: J66736.18
50 g, Each
Quantity
Catalog number: J66736.18
also known as J66736-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
56038-13-2
IUPAC Name
(2R,3R,4R,5R,6R)-2-{[(2R,3S,4S,5S)-2,5-bis(chloromethyl)-3,4-dihydroxyoxolan-2-yl]oxy}-5-chloro-6-(hydroxymethyl)oxane-3,4-diol
Molecular Formula
C12H19Cl3O8
InChI Key
BAQAVOSOZGMPRM-QBMZZYIRSA-N
SMILES
OC[C@H]1O[C@H](O[C@]2(CCl)O[C@H](CCl)[C@@H](O)[C@@H]2O)[C@H](O)[C@@H](O)[C@H]1Cl
Form
Crystals or powder or crystalline powder
Optical Rotation
+84 to +88? (c=1, water)
Identification (FTIR)
Conforms
Water Content (Karl Fischer Titration)
≤2%
Particle Size
≥95% is ≤100 mesh (≤149 microns)

Description

Sucralose is a non-caloric sugar used as a sweetener that activates T1R2/T1R3 sweet taste receptors and modulates glucagon-like peptide-1, glucose-dependent insulinotrophic peptide, and incretin secretion.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sucralose is a non-caloric sugar used as a sweetener that activates T1R2/T1R3 sweet taste receptors and modulates glucagon-like peptide-1, glucose-dependent insulinotrophic peptide, and incretin secretion.

Solubility
Soluble in Water.

Notes
Research use only: Not intended for food, cosmetic, or diagnostic and therapeutic use in animals nor humans.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text