[Glu1] Fibrinopeptide B, Human
[Glu<sup>1</sup>] Fibrinopeptide B, Human
Thermo Scientific Chemicals

[Glu1] Fibrinopeptide B, Human

 
Have Questions?
Quantity:
1 mg
Catalog number J66755.MCR
also known as J66755-M^
Price (USD)/ Each
124.00
-
Add to cart
Quantity:
1 mg
Request bulk or custom format
Price (USD)/ Each
124.00
Add to cart
[Glu1] Fibrinopeptide B, Human
Catalog numberJ66755.MCR
Price (USD)/ Each
124.00
-
Add to cart
Specifications
SpeciesHuman
Purification MethodHPLC
Common Name[Glu1] Fibrinopeptide B
FormatPowder
Molecular Weight (g/mol)1570.57
View more
[Glu^1] Fibrinopeptide B is used as a indicator of coagulation and fibrinolytic activity. And is useful for the tuning and calibration of electrospray ionization and MALDI mass spectrometers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only
Contents & Storage
-30°C to -10°C