Thermo Scientific Chemicals

Propidium iodide, Thermo Scientific Chemicals

Catalog number: J66764.MC
100 mg, Each
Thermo Scientific Chemicals

Propidium iodide, Thermo Scientific Chemicals

Catalog number: J66764.MC
100 mg, Each
Quantity
Catalog number: J66764.MC
also known as J66764-MC
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
25535-16-4
IUPAC Name
3,8-diamino-5-{3-[diethyl(methyl)azaniumyl]propyl}-6-phenylphenanthridin-5-ium diiodide
Molecular Formula
C27H34I2N4
InChI Key
XJMOSONTPMZWPB-UHFFFAOYSA-M
SMILES
[I-].[I-].CC[N+](C)(CC)CCC[N+]1=C(C2=CC=CC=C2)C2=CC(N)=CC=C2C2=CC=C(N)C=C12
Appearance (Color)
Dark red
Form
Powder or crystals or crystalline powder or flakes
Assay from Supplier's CofA
≥94.0% (HPLC)

Description

Propidium iodide is a fluorescent stain for nucleic acids. Cell membrane integrity excludes propidium iodide from staining viable and apoptotic cells. Propidium iodide may be used in flow cytometry to evaluate cell viability when used with other dyes that stain viable cells or cells that are early in the apoptosis process.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Propidium iodide is a fluorescent stain for nucleic acids. Cell membrane integrity excludes propidium iodide from staining viable and apoptotic cells. Propidium iodide may be used in flow cytometry to evaluate cell viability when us
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text