Gramicidin, 95%
Thermo Scientific Chemicals

Gramicidin, 95%

For cellular and molecular biology applications
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
100 mg
500 mg
Catalog number J66771.03
also known as J66771-03
Price (USD)/ Each
187.65
Online exclusive
208.00 
Save 20.35 (10%)
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
187.65
Online exclusive
208.00 
Save 20.35 (10%)
Add to cart
Gramicidin, 95%
Catalog numberJ66771.03
Price (USD)/ Each
187.65
Online exclusive
208.00 
Save 20.35 (10%)
-
Add to cart
SpecificationsSpecification SheetSpecification Sheet
Purification MethodHPLC
Purity95%
FormatPowder
For Use With (Application)Targets primarily the cell membrane of gram positive bacteria and shows little effect against gram negative bacteria
Molecular Weight (g/mol)1882.29
View more
Targets primarily the cell membrane of Gram-positive bacteria and shows little effect against Gram-negative bacteria. Gramicidin is typically used as a topical antimicrobial agent because it also causes hemolysis of red blood cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only
Contents & Storage
2°C to 8°C