Thermo Scientific Chemicals

Mecillinam, 90+%, Thermo Scientific Chemicals

Catalog number: J66774.MC
100 mg, Each
Thermo Scientific Chemicals

Mecillinam, 90+%, Thermo Scientific Chemicals

Catalog number: J66774.MC
100 mg, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: J66774.MC
also known as J66774-MC
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
32887-01-7
IUPAC Name
(2S,5R,6R)-6-{[(azepan-1-yl)methylidene]amino}-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid
Molecular Formula
C15H23N3O3S
InChI Key
BWWVAEOLVKTZFQ-NTZNESFSSA-N
SMILES
CC1(C)S[C@@H]2[C@H](N=CN3CCCCCC3)C(=O)N2[C@H]1C(O)=O
Purity
≥90.0%
Form
Powder
Appearance (Color)
White to off-white
Water Content
≤6.0%

Description

Effective against Gram-negative bacteria

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text