Neuropeptide Y, Human
Neuropeptide Y, Human
Thermo Scientific Chemicals

Neuropeptide Y, Human

For cellular and molecular biology applications
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 mg
0.5 mg
Catalog number J66797.MCR
also known as J66797-M^
Price (USD)/ Each
368.00
-
Add to cart
Quantity:
1 mg
Request bulk or custom format
Price (USD)/ Each
368.00
Add to cart
Neuropeptide Y, Human
Catalog numberJ66797.MCR
Price (USD)/ Each
368.00
-
Add to cart
Specifications
SpeciesHuman
Purification MethodHPLC
FormatSolid
Molecular Weight (g/mol)4271.74
Product TypeNeuropeptide Y, human
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Contents & Storage
-30°C to -10°C