Thermo Scientific Chemicals

Bialaphos sodium, 85%, Thermo Scientific Chemicals

Catalog number: J66802.MC
100 mg, Each
Thermo Scientific Chemicals

Bialaphos sodium, 85%, Thermo Scientific Chemicals

Catalog number: J66802.MC
100 mg, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: J66802.MC
also known as J66802-MC
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
71048-99-2
IUPAC Name
sodium 2-(2-{2-amino-4-[hydroxy(methyl)phosphoryl]butanamido}propanamido)propanoate
Molecular Formula
C11H21N3NaO6P
InChI Key
RTWIRLHWLMNVCC-UHFFFAOYNA-M
SMILES
[Na+].CC(NC(=O)C(C)NC(=O)C(N)CCP(C)(O)=O)C([O-])=O
Appearance (Color)
Pale orange
Form
Powder
Assay from Supplier's CofA
≥80.0%

Description

Bialaphos Sodium Salt is a tripeptide antibiotic that is employed in the transformation experiments of many species of plants that make use of the bar gene for selection. It is more effective than glufosinate in corn. In wheat, bialaphos was the most reliable selection criteria used to identify bar transformed plants.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description
• Bialaphos sodium consists of two L-alanyl units attached to L-phosphinothricyl
• Bialaphos is a bacterial metabolite and an antimicrobial
Application
• Bialaphos is commonly used for transformation experiments in some plant species as a selection agent
• Bialaphos sodium can be used as a standard for metabolite and environmental contaminant determination in laboratory settings
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text