Thermo Scientific Chemicals

Phenyl beta-D-glucopyranoside, 98%, Thermo Scientific Chemicals

Catalog number: J66885.06
5 g, Each
Thermo Scientific Chemicals

Phenyl beta-D-glucopyranoside, 98%, Thermo Scientific Chemicals

Catalog number: J66885.06
5 g, Each
Quantity
Catalog number: J66885.06
also known as J66885-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1464-44-4
IUPAC Name
(2R,3S,4S,5R,6S)-2-(hydroxymethyl)-6-phenoxyoxane-3,4,5-triol
Molecular Formula
C12H16O6
InChI Key
NEZJDVYDSZTRFS-WLEIBRHLNA-N
SMILES
OC[C@H]1O[C@@H](OC2=CC=CC=C2)[C@H](O)[C@@H](O)[C@@H]1O
Assay (HPLC)
98.0% min.
Optical Rotation
-73?2?C, c=1 in water 20?C
Melting Point
170-176?C

Description

A substrate used for the determination of D-glucosidase

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text