Thermo Scientific Chemicals

5-Keto-D-gluconic acid potassium salt, 98%, Thermo Scientific Chemicals

Catalog number: J66916.06
5 g, Each
Thermo Scientific Chemicals

5-Keto-D-gluconic acid potassium salt, 98%, Thermo Scientific Chemicals

Catalog number: J66916.06
5 g, Each
Quantity
Catalog number: J66916.06
also known as J66916-06
Price (USD)
Quantity
-
Have Questions?

Description

Used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as pharmaceutical intermediates.

Solubility
Soluble in water 50mg/ml

Notes
Store at -20°C. Incompatible with heat, oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text