Levofloxacin, 98%, Thermo Scientific Chemicals
Levofloxacin, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Levofloxacin, 98%, Thermo Scientific Chemicals

Broad-spectrum antibiotic used in pharmacokinetic , antibiotic resistance, and resistance prevention studies. Levofloxacin is active against Gram-positive and Gram-negative bacteria. It inhibits DNA gyrase (type II topoisomerase) and topoisomerase IV, thereby inhibiting cell division.
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J66943.03
also known as J66943-03
Price (USD)/ Each
24.90
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
24.90
Add to cart
Levofloxacin, 98%, Thermo Scientific Chemicals
Catalog numberJ66943.03
Price (USD)/ Each
24.90
-
Add to cart
Chemical Identifiers
CAS100986-85-4
IUPAC Namebis((2S)-7-fluoro-2-methyl-6-(4-methylpiperazin-1-yl)-10-oxo-4-oxa-1-azatricyclo[7.3.1.0⁵,¹³]trideca-5(13),6,8,11-tetraene-11-carboxylic acid) hydrate
Molecular FormulaC36H42F2N6O9
InChI KeySUIQUYDRLGGZOL-RCWTXCDDSA-N
SMILESO.C[C@H]1COC2=C3N1C=C(C(O)=O)C(=O)C3=CC(F)=C2N1CCN(C)CC1.C[C@H]1COC2=C3N1C=C(C(O)=O)C(=O)C3=CC(F)=C2N1CCN(C)CC1
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Residue on ignition≤0.2%
Appearance (Color)White to yellow
Assay (unspecified)≥97.5% (dry basis)
Impurity contentRelated substances: ≤ 0.5%
Levofloxacin is a broad-spectrum antibiotic used in pharmacokinetic , antibiotic resistance , and resistance prevention studies. Levofloxacin is active against Gram-positive and Gram-negative bacteria. It inhibits DNA gyrase (type II topoisomerase) and topoisomerase IV, thereby inhibiting cell division.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Levofloxacin is a broad-spectrum antibiotic used in pharmacokinetic , antibiotic resistance , and resistance prevention studies. Levofloxacin is active against Gram-positive and Gram-negative bacteria. It inhibits DNA gyrase (type II topoisomerase) and topoisomerase IV, thereby inhibiting cell division.

Solubility
Slightly soluble in water or methanol. Soluble in glacial acetic acid or dichloromethane

Notes
Research Use only: Not intended for animal or human diagnostic or therapeutic use.
RUO – Research Use Only