Thermo Scientific Chemicals

Levofloxacin, 98%, Thermo Scientific Chemicals

Catalog number: J66943.06
5 g, Each
Thermo Scientific Chemicals

Levofloxacin, 98%, Thermo Scientific Chemicals

Catalog number: J66943.06
5 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: J66943.06
also known as J66943-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
100986-85-4
IUPAC Name
bis((2S)-7-fluoro-2-methyl-6-(4-methylpiperazin-1-yl)-10-oxo-4-oxa-1-azatricyclo[7.3.1.0⁵,¹³]trideca-5(13),6,8,11-tetraene-11-carboxylic acid) hydrate
Molecular Formula
C36H42F2N6O9
InChI Key
SUIQUYDRLGGZOL-RCWTXCDDSA-N
SMILES
O.C[C@H]1COC2=C3N1C=C(C(O)=O)C(=O)C3=CC(F)=C2N1CCN(C)CC1.C[C@H]1COC2=C3N1C=C(C(O)=O)C(=O)C3=CC(F)=C2N1CCN(C)CC1
Form
Powder
Residue on ignition
≤0.2%
Appearance (Color)
White to yellow
Assay (unspecified)
≥97.5% (dry basis)
Impurity content
Related substances: ≤ 0.5%

Description

Levofloxacin is a broad-spectrum antibiotic used in pharmacokinetic , antibiotic resistance , and resistance prevention studies. Levofloxacin is active against Gram-positive and Gram-negative bacteria. It inhibits DNA gyrase (type II topoisomerase) and topoisomerase IV, thereby inhibiting cell division.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Levofloxacin is a broad-spectrum antibiotic used in pharmacokinetic , antibiotic resistance , and resistance prevention studies. Levofloxacin is active against Gram-positive and Gram-negative bacteria. It inhibits DNA gyrase (type II topoisomerase) and topoisomerase IV, thereby inhibiting cell division.

Solubility
Slightly soluble in water or methanol. Soluble in glacial acetic acid or dichloromethane

Notes
Research Use only: Not intended for animal or human diagnostic or therapeutic use.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text