Riboflavin-5'-phosphate sodium salt dihydrate, 73.0% min., Thermo Scientific Chemicals
Riboflavin-5'-phosphate sodium salt dihydrate, 73.0% min., Thermo Scientific Chemicals
Thermo Scientific Chemicals

Riboflavin-5'-phosphate sodium salt dihydrate, 73.0% min., Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
Catalog number J66949.18
also known as J66949-18
Price (USD)/ Each
163.00
-
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)/ Each
163.00
Add to cart
Riboflavin-5'-phosphate sodium salt dihydrate, 73.0% min., Thermo Scientific Chemicals
Catalog numberJ66949.18
Price (USD)/ Each
163.00
-
Add to cart
Chemical Identifiers
CAS6184-17-4
IUPAC Namesodium 10-[(2S,3S,4R)-5-(hydrogen phosphonatooxy)-2,3,4-trihydroxypentyl]-7,8-dimethyl-2H,3H,4H,10H-benzo[g]pteridine-2,4-dione dihydrate
Molecular FormulaC17H24N4NaO11P
InChI KeyLQVGUKOCMOKKJU-FTNKLFHDNA-M
SMILESO.O.[Na+].CC1=C(C)C=C2N(C[C@H](O)[C@H](O)[C@H](O)COP(O)([O-])=O)C3=NC(=O)NC(=O)C3=NC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Orange
Assay (UV/vis)73.0% min.
Optical Rotation+37 to +43? (1.5% dry w/v, 20%w/v HCl)
FormPowder
Loss on Drying< 10.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only