Endomorphin-2
Thermo Scientific Chemicals

Endomorphin-2

Have Questions?
Change viewbuttonViewtableView
Quantity:
25mg
5mg
Catalog number J66953.MB
also known as J66953-MB
Price (USD)/ Each
439.00
-
Add to cart
Quantity:
25mg
Request bulk or custom format
Price (USD)/ Each
439.00
Add to cart
Endomorphin-2
Catalog numberJ66953.MB
Price (USD)/ Each
439.00
-
Add to cart
Specifications
CAS141801-26-5
ColorWhite
FormatSolid
Molecular Weight (g/mol)571.678
Product TypeEndomorphin-2
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Contents & Storage
Store at -20°C