Thermo Scientific Chemicals

17-(Allylamino)-17-demethoxygeldanamycin, 99%, Thermo Scientific Chemicals

Catalog number: J66960.MB
25 mg, Each
Thermo Scientific Chemicals

17-(Allylamino)-17-demethoxygeldanamycin, 99%, Thermo Scientific Chemicals

Catalog number: J66960.MB
25 mg, Each
Quantity
Catalog number: J66960.MB
also known as J66960-MB
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
75747-14-7
IUPAC Name
(4Z,6E,8R,9R,10E,12R,13S,14R,16R)-13-hydroxy-8,14-dimethoxy-4,10,12,16-tetramethyl-3,20,22-trioxo-19-[(prop-2-en-1-yl)amino]-2-azabicyclo[16.3.1]docosa-1(21),4,6,10,18-pentaen-9-yl carbamate
Molecular Formula
C31H43N3O8
InChI Key
AYUNIORJHRXIBJ-CQFHKJEWSA-N
SMILES
CO[C@@H]1C[C@H](C)CC2=C(NCC=C)C(=O)C=C(NC(=O)C(C)=C/C=C/[C@@H](OC)[C@H](OC(N)=O)C(C)=C[C@@H](C)[C@@H]1O)C2=O
Appearance (Color)
Red to dark red
Assay from Supplier's CofA
≥98.5% (HPLC)
Form
Powder

Description

17-(Allylamino)-17-demethoxygeldanamycin (17-AAG) inhibits heat shock protein 90 (Hsp90), the expression of heat shock factor-1, and the activity of oncogenic proteins such as N-ras, Ki-ras, c-Akt, and p185erB2. In addition to its anti-tumor effects, 17-AAG also induces apoptosis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in DMSO at 150 mg/mL; soluble in ethanol at 5 mg/mL. Very poorly soluble in water

Notes
Research Use only: Not intended for animal or human diagnostic or therapeutic use.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text