Pullulan, Thermo Scientific Chemicals
Pullulan, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Pullulan, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
1 g
5 g
Catalog number J66961.09
also known as J66961-09
Price (USD)/ Each
123.65
Online exclusive
137.00 
Save 13.35 (10%)
-
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)/ Each
123.65
Online exclusive
137.00 
Save 13.35 (10%)
Add to cart
Pullulan, Thermo Scientific Chemicals
Catalog numberJ66961.09
Price (USD)/ Each
123.65
Online exclusive
137.00 
Save 13.35 (10%)
-
Add to cart
Chemical Identifiers
CAS9057-02-7
Molecular Formula(C18H30O15)AHO
SMILES*.O-*.[H][C@]1(-*)O[C@H](CO)[C@@H](O[C@@]2([H])O[C@H](CO)[C@@H](O[C@@]3([H])O[C@H](CO-*)[C@@H](O)[C@H](O)[C@H]3O)[C@H](O)[C@H]2O)[C@H](O)[C@H]1O
Molecular Weight (g/mol)NaN
SpecificationsSpecification SheetSpecification Sheet
Loss on Drying≤ 8%
Residue on ignition≤ 8%
FormWhite to off white crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only