[Leu13] Motilin, Human, Porcine
[Leu<sup>13</sup>] Motilin, Human, Porcine
Thermo Scientific Chemicals

[Leu13] Motilin, Human, Porcine

For cellular and molecular biology applications
Have Questions?
Quantity:
0.5 mg
Catalog number J66962.EXD
also known as J66962-!D
Price (USD)/ Each
191.00
-
Add to cart
Quantity:
0.5 mg
Request bulk or custom format
Price (USD)/ Each
191.00
Add to cart
[Leu13] Motilin, Human, Porcine
Catalog numberJ66962.EXD
Price (USD)/ Each
191.00
-
Add to cart
SpecificationsSpecification SheetSpecification Sheet
SpeciesHuman, Porcine
Purification MethodHPLC
FormatSolid
Molecular Weight (g/mol)2681.10
Product Type[Leu13] Motilin, human, porcine
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Contents & Storage
-30°C to -10°C