It is used and studied as a vasoprotective anti-inflammatory, anti-edematous and anti-nociceptive agent.1 It may be used to study its pharmacokinetics, safety and efficacy as a chemosensitizer and modulator of NF-κ B cell signaling
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used and studied as a vasoprotective anti-inflammatory, anti-edematous and anti-nociceptive agent.1 It may be used to study its pharmacokinetics, safety and efficacy as a chemosensitizer and modulator of NF-κ B cell signaling
Solubility
Soluble in methanol.
Notes
Desiccate at room temperature. Incompatible with oxidizing agents.
RUO – Research Use Only
Cesare R. Sirtori. Aescin: pharmacology, pharmacokinetics and therapeutic profile. Pharmacol. Res. 2001, 44 (3), 183-193 .
Ferruccio Berti.; Claudio Omini.; Daniela Longiave. The mode of action of Aescin and the release of prostaglandins. Prostaglandins. 1977, 14 (2), 241-249 .