Thermo Scientific Chemicals

1,3,4,6-Tetra-O-acetyl-beta-D-glucosamine hydrochloride

Catalog number: J66975.06
5 g, Each
Thermo Scientific Chemicals

1,3,4,6-Tetra-O-acetyl-beta-D-glucosamine hydrochloride

Catalog number: J66975.06
5 g, Each
Quantity
Catalog number: J66975.06
also known as J66975-06
Price (USD)
Quantity
-
Have Questions?

Description

NSC 82044 is a potential metabolic inhibitor of cellular-membrane glycoconjugates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
NSC 82044 is a potential metabolic inhibitor of cellular-membrane glycoconjugates.

Solubility
Soluble in water at 10mg/ml

Notes
Keep container tightly closed in a dry and well-ventilated place. Store in cool place.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text