1,3,4,6-Tetra-O-acetyl-beta-D-glucosamine hydrochloride
1,3,4,6-Tetra-O-acetyl-beta-D-glucosamine hydrochloride
Thermo Scientific Chemicals

1,3,4,6-Tetra-O-acetyl-beta-D-glucosamine hydrochloride

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
1 g
5 g
25 g
Catalog number J66975.18
also known as J66975-18
Price (USD)/ Each
773.00
-
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)/ Each
773.00
Add to cart
1,3,4,6-Tetra-O-acetyl-beta-D-glucosamine hydrochloride
Catalog numberJ66975.18
Price (USD)/ Each
773.00
-
Add to cart
Specifications
Chemical Name or Material1,3,4,6-Tetra-O-acetyl-beta-D-glucosamine hydrochloride
CAS10034-20-5
ColorBrown to White
Recommended StorageAmbient temperatures
RTECS NumberLZ6574000
View more
NSC 82044 is a potential metabolic inhibitor of cellular-membrane glycoconjugates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
NSC 82044 is a potential metabolic inhibitor of cellular-membrane glycoconjugates.

Solubility
Soluble in water at 10mg/ml

Notes
Keep container tightly closed in a dry and well-ventilated place. Store in cool place.
RUO – Research Use Only

General References:

  1. KK Barnes.; CK Mann. Electrochemical oxidation of primary aliphatic amines. J. Org. Chem. 1967, 32, (5), 1474-1479.
  2. D Horton.; WE Mast.; KD Philips. Specifically deuterated, acetylated derivatives of 2-amino-2-deoxy-D-glucose. Nuclear magnetic resonance studies on migration of acetyl groups. J. Org. Chem. 1967, 32, (5), 1471-1474.