Amylin, Human
Thermo Scientific Chemicals

Amylin, Human

 
Have Questions?
Quantity:
1 mg
Catalog number J66984.MCR
also known as J66984-M^
Price (USD)/ Each
468.00
-
Add to cart
Quantity:
1 mg
Request bulk or custom format
Price (USD)/ Each
468.00
Add to cart
Amylin, Human
Catalog numberJ66984.MCR
Price (USD)/ Each
468.00
-
Add to cart
Specifications
SpeciesHuman
Purification MethodHPLC
FormatSolid
Molecular Weight (g/mol)3903.27
Product TypeAmylin, human
View more
Amylin functions as part of the endocrine pancreas and contributes to glycemic control. It functions as a synergistic partner to insulin. The overall effect is to slow the rate of appearance (Ra) of glucose in the blood after eating.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only
Contents & Storage
-30°C to -10°C