Thermo Scientific Chemicals

D-Gulonic acid-1,4-lactone, 98%, Thermo Scientific Chemicals

Catalog number: J66987.14
25 g, Each
Thermo Scientific Chemicals

D-Gulonic acid-1,4-lactone, 98%, Thermo Scientific Chemicals

Catalog number: J66987.14
25 g, Each
Quantity
Catalog number: J66987.14
also known as J66987-14
Price (USD)
Quantity
-
Have Questions?

Description

D-Gulono-1,4-lactone is a hexanoic acid used as a starting material in EPC syntheses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-Gulono-1,4-lactone is a hexanoic acid used as a starting material in EPC syntheses.

Solubility
Soluble in water

Notes
Store at room temperature. Incompatible with heat, oxidizing agents. Ensure good ventilation.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text