Thermo Scientific Chemicals

Florfenicol, 98%, Thermo Scientific Chemicals

Catalog number: J66995.06
5 g, Each
Thermo Scientific Chemicals

Florfenicol, 98%, Thermo Scientific Chemicals

Catalog number: J66995.06
5 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: J66995.06
also known as J66995-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
73231-34-2
IUPAC Name
2,2-dichloro-N-[(1R,2S)-3-fluoro-1-hydroxy-1-(4-methanesulfonylphenyl)propan-2-yl]acetamide
Molecular Formula
C12H14Cl2FNO4S
InChI Key
AYIRNRDRBQJXIF-NXEZZACHSA-N
SMILES
CS(=O)(=O)C1=CC=C(C=C1)[C@@H](O)[C@@H](CF)NC(=O)C(Cl)Cl
Assay (HPLC)
98.0% min.
Melting Point
153-157 ?C

Description

A bacteriostatic antibiotic similar to chloramphenicol which inhibits bacterial protein synthesis

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • A bacteriostatic antibiotic similar to chloramphenicol which inhibits bacterial protein synthesis
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text