Gold nanoparticles, 40nm, supplied in 0.1 mg/ml sodium citrate with stabilizer, OD1, 530nm absorption, Thermo Scientific Chemicals
Catalog numberJ67001.AE
Price (USD)/ Each
278.00
-
Add to cart
Specifications
Chemical Name or MaterialGold nanoparticles
ColorPink to Red
Name Notesupplied in 0.1mg/mL sodium citrate with stabilizer, OD: 1, 530nm absorption
CAS7440-57-5
Storage Note 12° to 8°C
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.