Thermo Scientific Chemicals

C6 Ceramide

Catalog number: J67034.MA
10 mg, Each
Thermo Scientific Chemicals

C6 Ceramide

Catalog number: J67034.MA
10 mg, Each
Quantity
Catalog number: J67034.MA
also known as J67034-MA
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
124753-97-5
IUPAC Name
N-[(4E)-1,3-dihydroxyoctadec-4-en-2-yl]hexanamide
Molecular Formula
C24H47NO3
InChI Key
NPRJSFWNFTXXQC-VHEBQXMUNA-N
SMILES
CCCCCCCCCCCCC\C=C\C(O)C(CO)NC(=O)CCCCC
Infrared spectrum
Conforms
Purity
≥97.0% (HPLC)
Form
Solid

Description

C-6 ceramide is a cell-permeable analog of naturally occurring ceramides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text