Silver nanoparticles, 20nm, 0.02 mg/ml, supplied in 2mM sodium citrate, 405nm absorption, Thermo Scientific Chemicals
Silver nanoparticles, 20nm, 0.02 mg/ml, supplied in 2mM sodium citrate, 405nm absorption, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Silver nanoparticles, 20nm, 0.02 mg/ml, supplied in 2mM sodium citrate, 405nm absorption, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
100 mL
Catalog number J67067.AC
also known as J67067-AC
Price (USD)/ Each
77.65
Online exclusive
85.80 
Save 8.15 (10%)
-
Add to cart
Quantity:
25 mL
Request bulk or custom format
Price (USD)/ Each
77.65
Online exclusive
85.80 
Save 8.15 (10%)
Add to cart
Silver nanoparticles, 20nm, 0.02 mg/ml, supplied in 2mM sodium citrate, 405nm absorption, Thermo Scientific Chemicals
Catalog numberJ67067.AC
Price (USD)/ Each
77.65
Online exclusive
85.80 
Save 8.15 (10%)
-
Add to cart
Specifications
Chemical Name or MaterialSilver nanoparticles
ColorYellow
Name Note0.02mg/mL, supplied in 2mM sodium citrate, 405nm absorption
CAS7440-22-4
Storage Note 12° to 8°C
View more
Silver nanoparticles applications include molecular diagnostics and photonic devices, which take advantage of the novel optical properties of these nanomaterials. An increasingly common application is the use of silver nanoparticles for antimicrobial coatings, and many textiles, keyboards, wound dressings, and biomedical devices now contain silver nanoparticles that continuously release a low level of silver ions to provide protection against bacteria.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Silver nanoparticles applications include molecular diagnostics and photonic devices, which take advantage of the novel optical properties of these nanomaterials. An increasingly common application is the use of silver nanoparticles for antimicrobial coatings, and many textiles, keyboards, wound dressings, and biomedical devices now contain silver nanoparticles that continuously release a low level of silver ions to provide protection against bacteria.

Solubility
Fully miscible.

Notes
Particles per ml: 6.2E+11
RUO – Research Use Only

General References:

  1. JJ Mock.; M Barbic.; DR Smith.; DA Schultz. Shape effects in plasmon resonance of individual colloidal silver nanoparticles. J. Chem. Phys.2002, 116, (6), 6755-6777.
  2. S Link.; ZL Wang.; MA El-Sayed. Alloy formation of gold-silver nanoparticles and the dependence of the plasmon absorption on their composition. J. Phys. Chem. 1999, 103, (18), 3529-3533.