Dimethyloxaloylglycine, 98%, Thermo Scientific Chemicals
Dimethyloxaloylglycine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Dimethyloxaloylglycine, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 mg
50 mg
100 mg
Catalog number J67077.MA
also known as J67077-MA
Price (USD)/ Each
41.20
-
Add to cart
Quantity:
10 mg
Request bulk or custom format
Price (USD)/ Each
41.20
Add to cart
Dimethyloxaloylglycine, 98%, Thermo Scientific Chemicals
Catalog numberJ67077.MA
Price (USD)/ Each
41.20
-
Add to cart
Specifications
Chemical Name or MaterialDimethyloxaloylglycine
CAS89464-63-1
Health Hazard 1H302
Health Hazard 2GHS H Statement
H302
Harmful if swallowed.
Health Hazard 3P264b-P270-P301+P312-P330-P501c
View more
DMOG is a cell permeable prolyl-4-hydroxylase inhibitor that regulates hypoxia-inducible factor

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only