Thermo Scientific Chemicals
Dimethyloxaloylglycine, 98%, Thermo Scientific Chemicals
Catalog number: J67077.MC
100 mg, Each
Thermo Scientific Chemicals
Dimethyloxaloylglycine, 98%, Thermo Scientific Chemicals
Catalog number: J67077.MC
100 mg, Each
Quantity
Catalog number: J67077.MC
also known as J67077-MC
Price (USD)
Quantity
-
Have Questions?
Description
DMOG is a cell permeable prolyl-4-hydroxylase inhibitor that regulates hypoxia-inducible factor