Isotetrandrine, 98%
Thermo Scientific Chemicals

Isotetrandrine, 98%

 
Have Questions?
Quantity:
1 mg
Catalog number J67081.MCR
also known as J67081-M^
Price (USD)/ Each
-
Quantity:
1 mg
Request bulk or custom format
Specifications
Quantity1 mg
DescriptionWhite
Recommended StorageStore at -20°C
Assay98%
CAS477-57-6
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Contents & Storage
-30°C to -10°C