Silver nanoparticles, 10nm, 0.02 mg/ml, supplied in 2mM sodium citrate, 390-400nm absorption
Silver nanoparticles, 10nm, 0.02 mg/ml, supplied in 2mM sodium citrate, 390-400nm absorption
Silver nanoparticles, 10nm, 0.02 mg/ml, supplied in 2mM sodium citrate, 390-400nm absorption
Silver nanoparticles, 10nm, 0.02 mg/ml, supplied in 2mM sodium citrate, 390-400nm absorption
Thermo Scientific Chemicals

Silver nanoparticles, 10nm, 0.02 mg/ml, supplied in 2mM sodium citrate, 390-400nm absorption

CAS: 7440-22-4 | Ag | 107.87 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
100 mL
Catalog number J67111.AC
also known as J67111-AC
Price (USD)
88.80
Each
-
Add to cart
Quantity:
25 mL
Request bulk or custom format
Price (USD)
88.80
Each
Add to cart
Specifications
CAS7440-22-4
ChEBICHEBI:9141
Chemical Name or MaterialSilver nanoparticles
ColorYellow
IUPAC Namesilver
View more
Ask our AI about this Product
Silver nanoparticles applications include molecular diagnostics and photonic devices, which take advantage of the novel optical properties of these nanomaterials. An increasingly common application is the use of silver nanoparticles for antimicrobial coatings, and many textiles, keyboards, wound dressings, and biomedical devices now contain silver nanoparticles that continuously release a low level of silver ions to provide protection against bacteria.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Silver nanoparticles applications include molecular diagnostics and photonic devices, which take advantage of the novel optical properties of these nanomaterials. An increasingly common application is the use of silver nanoparticles for antimicrobial coatings, and many textiles, keyboards, wound dressings, and biomedical devices now contain silver nanoparticles that continuously release a low level of silver ions to provide protection against bacteria.

Solubility
Fully miscible in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Air sensitive. Handle and store under inert gas.
RUO – Research Use Only

General References:

  1. JJ Mock.; M Barbic.; DR Smith.; DA Schultz. Shape effects in plasmon resonance of individual colloidal silver nanoparticles. J. Chem. Phys.2002, 116, (6), 6755-6777.
  2. S Link.; ZL Wang.; MA El-Sayed. Alloy formation of gold-silver nanoparticles and the dependence of the plasmon absorption on their composition. J. Phys. Chem. 1999, 103, (18), 3529-3533.
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link