Thermo Scientific Chemicals
Gold nanoparticles, 50nm, supplied in 0.1mM PBS, 95%, OD1, 535nm absorption, Thermo Scientific Chemicals
Catalog number: J67157.AC
25 mL, Each
Thermo Scientific Chemicals
Gold nanoparticles, 50nm, supplied in 0.1mM PBS, 95%, OD1, 535nm absorption, Thermo Scientific Chemicals
Catalog number: J67157.AC
25 mL, Each
Quantity
Catalog number: J67157.AC
also known as J67157-AC
Price (USD)
Quantity
-
Have Questions?
Description
For conjugate development by passive adsorption of proteins and other ligands to the gold nanoparticle surface. Conjugates can be used in blotting, electron microscopy and immunohistochemistry applications.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • For conjugate development by passive adsorption of proteins and other ligands to the gold nanoparticle surface
  • Conjugates can be used in blotting, electron microscopy and immunohistochemistry applications