Thermo Scientific Chemicals

Actinomycin D (1 mg/ml, DMSO), Thermo Scientific Chemicals

Catalog number: J67160.XF
1 mL, Each
Thermo Scientific Chemicals

Actinomycin D (1 mg/ml, DMSO), Thermo Scientific Chemicals

Catalog number: J67160.XF
1 mL, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: J67160.XF
also known as J67160-XF
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
50-76-0
IUPAC Name
2-amino-4,6-dimethyl-3-oxo-N1,N9-bis[2,5,9-trimethyl-1,4,7,11,14-pentaoxo-6,13-bis(propan-2-yl)-hexadecahydro-1H-pyrrolo[2,1-i]1-oxa-4,7,10,13-tetraazacyclohexadecan-10-yl]-3H-phenoxazine-1,9-dicarboxamide
Molecular Formula
C62H86N12O16
InChI Key
RJURFGZVJUQBHK-UHFFFAOYNA-N
SMILES
CC(C)C1NC(=O)C(NC(=O)C2=C3N=C4C(OC3=C(C)C=C2)=C(C)C(=O)C(N)=C4C(=O)NC2C(C)OC(=O)C(C(C)C)N(C)C(=O)CN(C)C(=O)C3CCCN3C(=O)C(NC2=O)C(C)C)C(C)OC(=O)C(C(C)C)N(C)C(=O)CN(C)C(=O)C2CCCN2C1=O
Form
Liqiud
Comment
Actinomycin D: 1 mg/mL
Appearance (Color)
Clear Yellow to Orange to Dark Orange-Red

Description

Actinomycin D has a wide-ranging applications ranging from a selection agent in cell culture, use in studies of suppressing HIV-replication and programmed cell death of PC12 cells, and as an antibiotic in treatment of various malignant neoplasms, including Wilm?s tumor and the sarcomas.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Actinomycin D has a wide-ranging applications ranging from a selection agent in cell culture, use in studies of suppressing HIV-replication and programmed cell death of PC12 cells, and as an antibiotic in treatment of various malignant neoplasms, including Wilm′s tumor and the sarcomas.

Solubility
Soluble in acetonitrilie or acetone at 10 mg/mL and in DMSO at a minimum of 1 mg/mL, slightly soluble in water.

Notes
Research Use only: Not intended for animal or human diagnostic or therapeutic use.
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text