Assay from Suppliers CofASpecific biological assay: ≥2.0x10⁷ IU/mg
Appearance (Color)White
Molecular Weight17.5 ±1.8kDa
Assay from Suppliers CofA≥95.0% (RP-HPLC)
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.