Polymixin B sulfate, 50 mg/ml in distilled water, sterile-filtered
Polymixin B sulfate, 50 mg/ml in distilled water, sterile-filtered
Thermo Scientific Chemicals

Polymixin B sulfate, 50 mg/ml in distilled water, sterile-filtered

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 x 1mL
1mL
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J67163.8EQ
also known as J67163-8=
Price (USD)/ Each
307.00
-
Add to cart
Quantity:
5 x 1mL
Request bulk or custom format
Price (USD)/ Each
307.00
Add to cart
Polymixin B sulfate, 50 mg/ml in distilled water, sterile-filtered
Catalog numberJ67163.8EQ
Price (USD)/ Each
307.00
-
Add to cart
Specifications
CAS1405-20-5
FormatLiquid
Molecular FormulaC48H84N16O17S
MDL NumberMFCD00131991
Molecular Weight (g/mol)1301.571
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Contents & Storage
Store at -20°C