Silver nanoparticles, 30nm, 0.02 mg/ml, supplied in 2mM sodium citrate, 410nm absorption, Thermo Scientific Chemicals
Silver nanoparticles, 30nm, 0.02 mg/ml, supplied in 2mM sodium citrate, 410nm absorption, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Silver nanoparticles, 30nm, 0.02 mg/ml, supplied in 2mM sodium citrate, 410nm absorption, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
25 mL
Catalog number J67207.AE
also known as J67207-AE
Price (USD)/ Each
215.00
-
Add to cart
Quantity:
100 mL
Request bulk or custom format
Price (USD)/ Each
215.00
Add to cart
Silver nanoparticles, 30nm, 0.02 mg/ml, supplied in 2mM sodium citrate, 410nm absorption, Thermo Scientific Chemicals
Catalog numberJ67207.AE
Price (USD)/ Each
215.00
-
Add to cart
Specifications
Chemical Name or MaterialSilver nanoparticles
ColorYellow
Name Note0.02mg/mL, supplied in 2mM sodium citrate, 410nm absorption
CAS7440-22-4
Storage Note 12° to 8°C
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only