General Description
• Blasticidin S hydrochloride is an antibiotic that works by inhibiting peptide bond formation by the ribosome
Application
• Blasticidin S hydrochloride is a nucleoside antibiotic that inhibits protein synthesis in prokaryotic and eukaryotic cells
• It inhibits peptide bond formation by the ribosome
General References: