Streptomycin sulfate, 100 mg/ml in distilled water, sterile-filtered
Streptomycin sulfate, 100 mg/ml in distilled water, sterile-filtered
Thermo Scientific Chemicals

Streptomycin sulfate, 100 mg/ml in distilled water, sterile-filtered

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mL
100 mL
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J67224.AD
also known as J67224-AD
Price (USD)/ Each
86.50
-
Add to cart
Quantity:
50 mL
Request bulk or custom format
Price (USD)/ Each
86.50
Add to cart
Streptomycin sulfate, 100 mg/ml in distilled water, sterile-filtered
Catalog numberJ67224.AD
Price (USD)/ Each
86.50
-
Add to cart
Specifications
Chemical Name or MaterialStreptomycin sulfate
CAS3810-74-0
Health Hazard 1H317-H361d
Health Hazard 3P201-P202-P261-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P501c
Recommended StorageStore at -20°C
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only