Clozapine N-oxide, 99%, Thermo Scientific Chemicals
Clozapine N-oxide, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Clozapine N-oxide, 99%, Thermo Scientific Chemicals

 
Have Questions?
Quantity:
5 mg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J67235.LB0
also known as J67235-#0
Price (USD)/ Each
186.00
-
Add to cart
Quantity:
5 mg
Request bulk or custom format
Price (USD)/ Each
186.00
Add to cart
Clozapine N-oxide, 99%, Thermo Scientific Chemicals
Catalog numberJ67235.LB0
Price (USD)/ Each
186.00
-
Add to cart
Specifications
Chemical Name or MaterialClozapine N-oxide
CAS34233-69-7
Health Hazard 1H301-H315-H319-H335
Health Hazard 2GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only