Thermo Scientific Chemicals

Puromycin dihydrochloride, 10 mg/ml in distilled water, sterile-filtered, Thermo Scientific Chemicals

Catalog number: J67236.XF
1 mL, Each
Thermo Scientific Chemicals

Puromycin dihydrochloride, 10 mg/ml in distilled water, sterile-filtered, Thermo Scientific Chemicals

Catalog number: J67236.XF
1 mL, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: J67236.XF
also known as J67236-XF
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
53-79-2
IUPAC Name
(2S)-2-amino-N-[(2S,4R,5R)-5-[6-(dimethylamino)-9H-purin-9-yl]-4-hydroxy-2-(hydroxymethyl)oxolan-3-yl]-3-(4-methoxyphenyl)propanamide
Molecular Formula
C22H29N7O5
InChI Key
RXWNCPJZOCPEPQ-WABMVTPESA-N
SMILES
COC1=CC=C(C[C@H](N)C(=O)NC2[C@@H](CO)O[C@H]([C@@H]2O)N2C=NC3=C(N=CN=C23)N(C)C)C=C1
Appearance (Color)
Clear colorless to pale cream
Assay (special)
Absorbance at 275nm (0.1mg/ml): 0.28 to 0.38
pH
6.0 to 7.0
Assay (special)
Bacterial endotoxin: ≤5.0 EU/mg
Form
Liquid

Description

Puromycin is also used as a substrate of Mdr-1. It is been widely used in cell culture applications. It also induces apoptosis which was found as parallel apoptosis caused by deprivation of neuronal growth factor (NGF) in developing chick sympathetic neurons; however, this apoptotic activity can be inhibited by several transcriptional inhibitors such as: actinomycin D and α-amanitin (sc-202440) which has lead researchers to hypothesize that apoptosis is triggered through the transcription of particular genes. Shown to arrest cells in G2/M phase. Puromycin is an inhibitor of CD13 and DPP7.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Puromycin (puromycine) is an aminoglycoside antibiotic originating from Streptomyces alboniger
  • It acts as an aminoacyl tRNA analog and prematurely terminates peptide elongation
  • Puromycin has effects on both prokaryotes and eukaryotes
  • Puromycin may also have neoplastic activity

Application

  • Puromycin can be used for research use as an immunofluorescent target, in antibiotic resistance testing, or for studies on drug mechanisms
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text