Gold nanoparticles, 10nm, supplied in 0.1mM PBS, reactant-free, 99%, OD1, 520nm absorption, Thermo Scientific Chemicals
Gold nanoparticles, 10nm, supplied in 0.1mM PBS, reactant-free, 99%, OD1, 520nm absorption, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Gold nanoparticles, 10nm, supplied in 0.1mM PBS, reactant-free, 99%, OD1, 520nm absorption, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
100 mL
Catalog number J67238.AC
also known as J67238-AC
Price (USD)/ Each
116.00
-
Add to cart
Quantity:
25 mL
Request bulk or custom format
Price (USD)/ Each
116.00
Add to cart
Gold nanoparticles, 10nm, supplied in 0.1mM PBS, reactant-free, 99%, OD1, 520nm absorption, Thermo Scientific Chemicals
Catalog numberJ67238.AC
Price (USD)/ Each
116.00
-
Add to cart
Specifications
Chemical Name or MaterialGold nanoparticles
ColorRed
Name Notesupplied in 0.1mM PBS, reactant-free, 99%, OD1, 520nm absorption
CAS7440-57-5
Storage Note 12° to 8°C
View more
Ideal for sensitive applications requiring a minimum amount of contaminating reactants. Well suited for protein conjugate development by passive adsorption to the gold nanoparticle surface. Conjugates can be used in blotting, electron microscopy and immunohistochemistry applications

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only