CommentAll components are dissolved in UV treated 18.2 megohm-cm ± 1 water, low endotoxin water, followed by pH adjustment and filtered through 0.22 micron filter
pH5.0 ± 0.15
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.