Vancomycin hydrochloride, 50 mg/ml in distilled water, sterile-filtered, Thermo Scientific Chemicals
Vancomycin hydrochloride, 50 mg/ml in distilled water, sterile-filtered, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Vancomycin hydrochloride, 50 mg/ml in distilled water, sterile-filtered, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 mL
5 x 1 mL
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J67251.XF
also known as J67251-XF
Price (USD)/ Each
89.30
-
Add to cart
Quantity:
1 mL
Request bulk or custom format
Price (USD)/ Each
89.30
Add to cart
Vancomycin hydrochloride, 50 mg/ml in distilled water, sterile-filtered, Thermo Scientific Chemicals
Catalog numberJ67251.XF
Price (USD)/ Each
89.30
-
Add to cart
Specifications
Chemical Name or MaterialVancomycin hydrochloride
TypeVancomycin Hydrochloride
Physical FormLiquid
CAS1404-93-9
Health Hazard 1H317-H335
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only