MES, 0.2M buffer soln., pH 6.0, low endotoxin
MES, 0.2M buffer soln., pH 6.0, low endotoxin
Thermo Scientific Chemicals

MES, 0.2M buffer soln., pH 6.0, low endotoxin

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
100 mL
Catalog number J67264.AK
also known as J67264-AK
Price (USD)/ Each
109.65
Online exclusive
122.00 
Save 12.35 (10%)
-
Add to cart
Quantity:
250 mL
Request bulk or custom format
Price (USD)/ Each
109.65
Online exclusive
122.00 
Save 12.35 (10%)
Add to cart
MES, 0.2M buffer soln., pH 6.0, low endotoxin
Catalog numberJ67264.AK
Price (USD)/ Each
109.65
Online exclusive
122.00 
Save 12.35 (10%)
-
Add to cart
Specifications
Chemical Name or MaterialMES
Name Notelow endotoxin
CAS145224-94-8
ColorColorless
Recommended StorageAmbient temperatures
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only