Thermo Scientific Chemicals

Gold nanoparticles, 20nm, supplied in 0.1 mg/ml sodium citrate with stabilizer, OD1, 524nm absorption, Thermo Scientific Chemicals

Catalog number: J67269.AC
25 mL, Each
Thermo Scientific Chemicals

Gold nanoparticles, 20nm, supplied in 0.1 mg/ml sodium citrate with stabilizer, OD1, 524nm absorption, Thermo Scientific Chemicals

Catalog number: J67269.AC
25 mL, Each
Quantity
Catalog number: J67269.AC
also known as J67269-AC
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7440-57-5
IUPAC Name
gold
Molecular Formula
Au
InChI Key
PCHJSUWPFVWCPO-UHFFFAOYSA-N
SMILES
[Au]
Form
Liquid
Concentration
1.00-1.05 (OD)
Polydispersity Index
≤0.2
Particle Size
Mean Hydrodynamic Diameter: 28.0-32.0 nm
Particle Size
Core Size: 18.0-22.0 nm

Description

Colloidal gold is an electron-dense, non-fading marker useful as a probe in electron microscopy, light microscopy and immunoblotting. It requires no additional processing for detection, but in some applications the signal can be enhanced by reaction with silver. It can be complexed with biomolecules by strong, non-covalent interactions. Particles less than 15 nm are most useful in transmission electron microscopy and for applications where access to the probe may be hindered by larger particles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Colloidal gold is an electron-dense, non-fading marker useful as a probe in electron microscopy, light microscopy and immunoblotting. It requires no additional processing for detection, but in some applications the signal can be enhanced by reaction with silver. It can be complexed with biomolecules by strong, non-covalent interactions. Particles less than 15 nm are most useful in transmission electron microscopy and for applications where access to the probe may be hindered by larger particles.

Solubility
Fully miscible in water. It is soluble in aqua regia, nitric acid and hydrochloric acid.

Notes
Particles per ml: 6.54E+11
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text